• USA Home
  • A8303 - Albumin, α-D-mannopyranosylphenyl isothiocyanate bovine

EMAIL THIS PAGE TO A FRIEND
A8303 Sigma

Albumin, α-D-mannopyranosylphenyl isothiocyanate bovine

lyophilized powder

Synonym: α-D-Mannosylated-PITC-albumin

Purchase

Properties

Related Categories Albumin, Biochemicals and Reagents, Control Proteins for Glycosylation Studies, Glycobiology, Modified Albumins,
form   lyophilized powder
extent of labeling   15-25 mol α-D-mannopyranose per mol albumin
storage temp.   −20°C

Description

Application

Mannosylated BSA was used to assess the feasibility of using mannosylated proteins in general as liver-specific delivery systems. Mannosylation was found to confer a greater liver bioaccumulation rate than galactosylation, succinylation, or cationic modification.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?