• USA Home
  • A8335 - Albumin, bovine-β-melibiosyl-1-N-glycyl-malonyl

EMAIL THIS PAGE TO A FRIEND
A8335 Sigma

Albumin, bovine-β-melibiosyl-1-N-glycyl-malonyl

Synonym: α-D-Gal-(1→6)-β-D-Glc-1→N-(Gly-Malonyl-albumin

Purchase

Properties

storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?