• USA Home
  • A8342 - Mild Alkaline Northern Wash solution
EMAIL THIS PAGE TO A FRIEND
A8342 Sigma
Mild Alkaline Northern Wash solution
Purchase
Description
Application
Mild alkaline Northern wash solution is used to wash the solid support (e.g. nylon membrane) following capillary transfer of RNA using the alkaline Northern blotting procedure.
Other Notes
2× SSC, 0.1% SDS
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?