• USA Home
  • A8435 - Alcohol Dehydrogenase, NADP+ dependent from Thermoanaerobium brockii

EMAIL THIS PAGE TO A FRIEND
A8435 Sigma

Alcohol Dehydrogenase, NADP+ dependent from Thermoanaerobium brockii Green Alternative

lyophilized powder, 5-15 units/mg protein

Synonym: Alcohol:NADP+ oxidoreductase, TBADH

Purchase

Properties

Related Categories 1.1.x.x Acting on hydroxyl groups, 1.x.x.x Oxidoreductases, Alcohol Metabolism, Application Index, Biochemicals and Reagents,
form   lyophilized powder
composition   Protein, ≥20% biuret
greener alternative product characteristics   Waste Prevention: Greener alternative product characteristics
Learn more about the Principles of Green Chemistry.
greener alternative category   Enabling
storage temp.   −20°C

Description

Biochem/physiol Actions

An extremely thermostable enzyme with broad substrate specificity for alcohols, ketones and acetaldehyde.

Alcohol Dehydrogenase, from Thermoanaerobium brockii, is an extremely thermostable enzyme with broad substrate specificity for alcohols, ketones and acetaldehyde. Alcohol dehydrogenases (ADHs) are enzymes that catalyze the reversible conversion of ketone/aldehydes to the corresponding alcohols using a nicotinamide cofactor.

General description

Sigma Life Science is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for waste prevention when used in fuel cell research. For more information see the article in biofiles.

Unit Definition

One unit will oxidize 1.0 μmole of 2-propanol to acetone per min at pH 7.8 at 40 °C in the presence of NADP+.

Physical form

Contains phosphate buffer salts and dithioerythritol

Application

Alcohol dehydrogenase may be used to synthesize enantiomerically pure stereoisomers of chiral alcohols. It may be used to study ethanol fuel cells, alcoholism and drug dependence. Product A8435 is NADP+ dependent and is obtained from Thermoanaerobium brockii. It contains phosphate buffer salts and dithioerythritol. The product has been assayed with 2-propanol and NADP+ to analyse the function of fatty aldehyde reductase (FALDR) derived from Marinobacter aquaeolei VT8.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?