• USA Home
  • A8529 - Albumin from chicken egg white

EMAIL THIS PAGE TO A FRIEND
A8529 Sigma

Albumin from chicken egg white

suitable for marker for electrophoresis (non-denaturing PAGE)

Synonym: Albumin from hen egg white, Ovalbumin

Purchase

Properties

Related Categories Albumin, Albumins by Application, Albumins from other species, Biochemicals and Reagents, For Molecular Weight Markers and Protein Standards,
form   crystalline
mol wt   mol wt 44287 Da by calculation
  mol wt ~45 kDa
solubility   H2O: soluble2 mg + 1  mL, clear, colorless
suitability   suitable for marker for electrophoresis (non-denaturing PAGE)
storage temp.   −20°C

Description

Application

A phosphorylated glycoprotein which is useful as a marker (~45 kDa) for protein electrophoresis. Has been used to produce an animal model of asthma.

Reconstitution

0.9-1.4 mg/mL after reconstitution with 1 mL of water

Protocols & Applications

Select the right albumin for your application with the Albumin Selection Guide

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
AY9384000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

MWND500 Kit for Molecular Weights 14,000-500,000 Non-denaturing
T9767 Trypsin inhibitor from Glycine max (soybean), BioReagent, For use as a marker in SDS-PAGE

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?