• USA Home
  • A8547 - 5-Androstene-3β,17β-diol 3-sulfate triethylammonium salt

EMAIL THIS PAGE TO A FRIEND
A8547 Sigma

5-Androstene-3β,17β-diol 3-sulfate triethylammonium salt

Purchase

Properties

form   neat
drug control   USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?