• USA Home
  • A8699 - Activin Receptor IIA human

EMAIL THIS PAGE TO A FRIEND
A8699 Sigma

Activin Receptor IIA human

>95% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder

Purchase

Properties

Related Categories Activins, Activins and Inhibins, Cell Biology, Cell Signaling and Neuroscience, Cytokines and Growth Factors,
recombinant   expressed in baculovirus infected Sf21 cells
assay   >95% (SDS-PAGE)
form   lyophilized powder
impurities   endotoxin, tested
storage temp.   −20°C
Gene Information   human ... ACVR2A(92)

Description

Analysis Note

The biological activity is measured by its ability to inhibit hemoglobin expression induced by activin in K562 cells.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5 mg bovine serum albumin.

Biochem/physiol Actions

Activin isoforms are members of the TGF-β superfamily that bind to heteromeric complexes of a type I and a type II serine/threonine kinase receptor, both of which are essential for signal transduction. In addition to the activin isoforms, activin receptor II binds inhibin, BMP-2 and BMP-7. Human, murine and rat type II activin receptors demonstrate greater than 98% homology.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?