EMAIL THIS PAGE TO A FRIEND
A8723 Sigma

Altretamine

solid

Synonym: N,N,N′,N′,N′′,N′′-Hexamethyl-1,3,5-triazine-2,4,6-triamine

  • CAS Number 645-05-6

  • Empirical Formula (Hill Notation) C9H18N6

  • Molecular Weight 210.28

  •  MDL number MFCD00549245

  •  PubChem Substance ID 24278242

Purchase

Description

Biochem/physiol Actions

Antineoplastic

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?