EMAIL THIS PAGE TO A FRIEND
A8785 Sigma

Alkali Blue 6B

Synonym: Acid Blue 119

  •  Colour Index Number 42765

  •  EC Number 232-331-3

Purchase

Properties

composition   Dye content, ~85%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?