• USA Home
  • A8827 - Adenosine 5′-triphosphate calcium salt

EMAIL THIS PAGE TO A FRIEND
A8827 Sigma

Adenosine 5′-triphosphate calcium salt

90-95%

Purchase

Properties

assay   90-95%
solubility   H2O: 50 mg/mL
  H2O: soluble
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

A2383 Adenosine 5′-triphosphate disodium salt hydrate, Grade I, ≥99%, from microbial

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?