• USA Home
  • A9030 - 5β-Androst-9(11)-en-3α-ol-17-one

EMAIL THIS PAGE TO A FRIEND
A9030 Sigma

5β-Androst-9(11)-en-3α-ol-17-one

Synonym: 3α-Hydroxy-5β,9(11)-androsten-17-one

Purchase

Properties

drug control   regulated under CDSA - not available from Sigma-Aldrich Canada

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?