• USA Home
  • A9153 - Acetic acid N-hydroxysuccinimide ester

EMAIL THIS PAGE TO A FRIEND
A9153 Sigma

Acetic acid N-hydroxysuccinimide ester

≥95% (TLC)

Purchase

Properties

assay   ≥95% (TLC)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?