• USA Home
  • A9290 - N-Acetylneuraminic acid

EMAIL THIS PAGE TO A FRIEND
A9290 Sigma

N-Acetylneuraminic acid

cell culture tested, ≥95%

Synonym: 5-Acetamido-3,5-dideoxy-D-glycero-D-galactononulosonic acid, Lactaminic acid, NAN, NANA, Sialic acid

  • CAS Number 131-48-6

  • Empirical Formula (Hill Notation) C11H19NO9

  • Molecular Weight 309.27

  •  Beilstein Registry Number 1716283

  •  EC Number 205-023-1

  •  MDL number MFCD00006620

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   SQVRNKJHWKZAKO-PFQGKNLYSA-N
assay   ≥95%
suitability   cell culture tested
storage temp.   −20°C

Description

Biochem/physiol Actions

Both sialic acid and neuraminic acid are loosely used to refer to conjugates of neuraminic acid. N-Acetylneuraminic acid is often found as the terminal sugar of cell surface glycoproteins. Cell surface glycoproteins have important roles in cell recognition and interaction as well as in cell adhesion. Membrane glycoproteins are also important in tumor growth and metastases.

Other Notes

Synthetic

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

A0812 N-Acetylneuraminic acid, synthetic, ≥95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?