• USA Home
  • A9326 - 1-(4,5-Dimethoxy-2-nitrophenyl)ethyl arachidonate

EMAIL THIS PAGE TO A FRIEND
A9326 Sigma

1-(4,5-Dimethoxy-2-nitrophenyl)ethyl arachidonate

≥95%

Synonym: Arachidonic acid 1-(4,5-dimethoxy-2-nitrophenyl)ethyl ester

Purchase

Properties

InChI Key   LIKGQASPYMHUMB-SNPVRQPZSA-N
assay   ≥95%
storage temp.   −20°C

Description

Application

Photoactive probe for arachidonic acid receptors.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?