• USA Home
  • A9329 - Aldolase from spinach

EMAIL THIS PAGE TO A FRIEND
A9329 Sigma

Aldolase from spinach

lyophilized powder, 0.3-1.0 units/mg protein

Synonym: D-Fructose-1,6-bisphosphate-D-glyceraldehyde-3-phosphate-lyase

Purchase

Properties

Related Categories 4.1.x.x C-C bonds, 4.x.x.x Lyases, Biochemicals and Reagents, Enzyme Class Index, Enzymes, Inhibitors, and Substrates More...
form   lyophilized powder
shipped in   dry ice
storage temp.   −20°C

Description

Analysis Note

Protein determined by Lowry

Quality

Crude

Unit Definition

One unit will convert 1.0 μmole of fructose 1,6-diphosphate to dihydroxyacetone phosphate and glyceraldehyde 3-phosphate per min at pH 7.4 at 25 °C.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?