• USA Home
  • A9398 - Arg-Arg 4-methoxy-β-naphthylamide trihydrochloride

EMAIL THIS PAGE TO A FRIEND
A9398 Sigma

Arg-Arg 4-methoxy-β-naphthylamide trihydrochloride

  • CAS Number 42761-77-3

  • Empirical Formula (Hill Notation) C23H35N9O3 · 3HCl

  • Molecular Weight 594.97

  •  MDL number MFCD00058036

  •  PubChem Substance ID 24891444

Purchase

Description

Substrates

Fluorogenic substrate for dipeptidyl aminopeptidase III.

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?