EMAIL THIS PAGE TO A FRIEND
A9436 Sigma

N-Acetyl-β-Asp-Glu

  • CAS Number 4910-46-7

  • Empirical Formula (Hill Notation) C11H16N2O8

  • Molecular Weight 304.25

  •  MDL number MFCD00133559

  •  PubChem Substance ID 24891453

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   GUCKKCMJTSNWCU-UHFFFAOYSA-N
storage temp.   −20°C

Description

Amino Acid Sequence

Nac-Asp-Glu

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Biochem/physiol Actions

N-Acetyl-β-Asp-Glu (βNAAG) is used as a substrate/inhibitor to identify and characterize glutamate carboxypeptidase(s). βNAAG is often used together witn N-Acetyl-Asp-Glu (NAAG), a neuropeptide, to differentiate the activity of glutamate carboxypeptidase activity and function in vivo.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?