• USA Home
  • A9471 - Acetyl-Arg-Tyr-Tyr-Arg-Ile-Lys Amide

EMAIL THIS PAGE TO A FRIEND
A9471 Sigma

Acetyl-Arg-Tyr-Tyr-Arg-Ile-Lys Amide

>97% (HPLC), solid

Synonym: Ac-RYYRIK-NH2

  • Empirical Formula (Hill Notation) C44H70N14O9

  • Molecular Weight 939.12

  •  MDL number MFCD03452829

Purchase

Description

Amino Acid Sequence

Ac-Arg-Tyr-Tyr-Arg-Ile-Lys-NH2

Application

Competitive mixed ORL-1 nociceptin/orphanin FQ receptor agonist/antagonist.

Price and Availability


Better Together

Discover your Target

Making Headway
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?