• USA Home
  • A9527 - ε-Aminocaproyl-p-chlorobenzylamide hydrochloride

EMAIL THIS PAGE TO A FRIEND
A9527 Sigma

ε-Aminocaproyl-p-chlorobenzylamide hydrochloride

  • CAS Number 73655-06-8

  • Empirical Formula (Hill Notation) C13H19ClN2O · HCl

  • Molecular Weight 291.22

  •  MDL number MFCD00039070

  •  PubChem Substance ID 24891467

Purchase

Properties

Related Categories Amides, Building Blocks, C8 to C20+, Carbonyl Compounds, Chemical Synthesis,
InChI Key   VJZWAGBAIKLRNF-UHFFFAOYSA-N
storage temp.   2-8°C

Description

Application

Ligand for affinity purification of thrombin and prekallikrein.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?