• USA Home
  • A9577 - 3-Amino-2,5-dichlorobenzoic acid

EMAIL THIS PAGE TO A FRIEND
A9577 Sigma

3-Amino-2,5-dichlorobenzoic acid

≥97%

Purchase

Properties

assay   ≥97%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
DG1925000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?