EMAIL THIS PAGE TO A FRIEND
A9580 Sigma

p-Aminobenzoic acid

cell culture tested, insect cell culture tested, ≥99%

Synonym: PABA

Purchase

Properties

assay   ≥99%
suitability   cell culture tested
  insect cell culture tested

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
DG1400000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

A9878 4-Aminobenzoic acid, ReagentPlus®, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?