• USA Home
  • A9590 - AP-1 Consensus Oligonucleotide

EMAIL THIS PAGE TO A FRIEND
A9590 Sigma

AP-1 Consensus Oligonucleotide

Synonym: 5′-CGC TTG ATG ACT CAG CCG GAA-3′

Purchase

Description

Application

Supplied lyophilized; after reconstitution, can be end-labeled with polynucleotide kinase.

Other Notes

Double-stranded oligonucleotide representing the binding sequence of the AP-1 transcription factors (homo- and heterodimers of the Jun and Fos family of proteins).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?