• USA Home
  • A9685 - (S)-Aromatic Alcohol Dehydrogenase, NADP+ dependent from Thermoanaerobium sp.

EMAIL THIS PAGE TO A FRIEND
A9685 Sigma

(S)-Aromatic Alcohol Dehydrogenase, NADP+ dependent from Thermoanaerobium sp.

lyophilized powder, 1-5 units/mg solid

Synonym: Alcohol:NADP+ oxidoreductase

Purchase

Description

Application

A thermostable enzyme with broad specificity for alcohols and ketones. Highly enantioselective for many (S)-aromatic alcohols. Chiral selectivity Vs/Vr ≥40 using (S)- and (R)-sec-phenethyl alcohol.

Unit Definition

One unit will oxidize 1.0 μmole of 2-propanol to acetone per min at pH 9.0 at 50 °C.

Physical form

Lyophilized powder containing raffinose, dithiothreitol, and potassium phosphate buffer salt.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?