• USA Home
  • A9701 - Acetate Kinase from Methanosarcina thermophila

EMAIL THIS PAGE TO A FRIEND
A9701 Sigma

Acetate Kinase from Methanosarcina thermophila

recombinant, expressed in E. coli, lyophilized powder, ≥800 units/mg protein (Bradford)

Synonym: ATP:Acetate phosphotransferase

Purchase

Description

Unit Definition

One unit will phosphorylate 1.0 μmole of acetate to acetyl phosphate per min at pH 7.6 at 25 °C.

Physical form

Lyophilized powder containing Tris buffer salt, KCl, DTT and Ficoll®.

Legal Information

Ficoll is a registered trademark of GE Healthcare

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

inhibitor

Product #

Description

Add to Cart

D5520 Dimethyl dicarbonate, for histidine methylation

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?