• USA Home
  • A9721 - Adhesion Molecule Factsbook, 2nd ed.

EMAIL THIS PAGE TO A FRIEND
A9721 Sigma

Adhesion Molecule Factsbook, 2nd ed.

  •  ISBN-10 0-12-356505-7

  •  ISBN-13 978-0-12-356505-1

Purchase

Properties

Related Categories Biochemistry Books, Books and Software, Labware More...
publication info   C. Isacke and M. Horton, Academic Press, 2000, 313 pp., soft cover
mfr. no.   Elsevier

Description

General description

Second Edition is completely updated, and expanded to reflect new advances. It contains over 80 entries on all currently cloned and characterized cell adhesion molecules, including the families of integrins, cadherins, selectins, members of the immunoglobulin superfamily, and other relevant receptors. It provides information on amino acid sequence of the most commonly studied organisms; gene organization and chromosomal location; alternative nomenclature; biological function; tissue distribution; disease association; database accession numbers.

Table of Contents

Topics include
• Protein structure and molecular weights
• Amino acid sequence of the most commonly studied organisms
• Gene organization and chromosomal location
• Alternative nomenclature
• Ligands
• Biological function
• Tissue distribution
• Disease association
• Knockout mice
• Database accession numbers
• How to use access info on the Web and Web links

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?