EMAIL THIS PAGE TO A FRIEND
A9750 Sigma

3-Acetylindole

practical grade

Synonym: 3-Indolyl methyl ketone, NSC 47180, NSC 58084

  • CAS Number 703-80-0

  • Empirical Formula (Hill Notation) C10H9NO

  • Molecular Weight 159.18

  •  Beilstein Registry Number 122579

  •  EC Number 211-875-5

  •  MDL number MFCD00005626

  •  PubChem Substance ID 24891491

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Building Blocks, C10, Chemical Synthesis, Heterocyclic Building Blocks, Indoles More...
grade   practical grade
InChI Key   VUIMBZIZZFSQEE-UHFFFAOYSA-N
mp   188-192 °C(lit.)

Description

Application

Reactant for preparation of:
• Indole derivatives as antitumor agents
• Endothelin-1 antagonists
• Oncrasin-1 derivatives as inhibitors of the C-terminal domain of RNA polymerase II with antitumor activities
• Inhibitors of hepatitis C NS3/4A serine protease
• Antibacterial agens against MDR Staphylococcus aureus strains
• Antimalarial agents
• Anti-bovine viral diarrhea virus (BVDV) agents
• HIV-1 integrase inhibitors
• Inhibitors of NF-κB transcription regulation related to TNF-α cytokine release

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
OB4520000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?