EMAIL THIS PAGE TO A FRIEND
A9790 Sigma

Adonitol

BioReagent, suitable for cell culture

Synonym: Adonite, Ribitol

  • CAS Number 488-81-3

  • Empirical Formula (Hill Notation) C5H12O5

  • Molecular Weight 152.15

  •  Beilstein Registry Number 1720524

  •  EC Number 207-685-7

  •  MDL number MFCD00064291

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Cell Culture, Miscellaneous Reagents and Supplements, Reagents and Supplements More...
InChI Key   HEBKCHPVOIAQTA-ZXFHETKHSA-N
product line   BioReagent
suitability   suitable for cell culture

Description

Application

Useful for studies of the thermo-tolerance of cells in cell culture.

Price and Availability


Filter out uncertainty.

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?