• USA Home
  • A9811 - Alkaline Phosphatase-Anti-Alkaline Phosphatase antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
A9811 Sigma

Alkaline Phosphatase-Anti-Alkaline Phosphatase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: APAAP

Purchase

Properties

Related Categories AL-AL, APAAP, Alphabetical Index, Antibodies, Primary Antibodies,
biological source   rabbit
antibody form   affinity isolated antibody
form   buffered aqueous solution
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:10
shipped in   wet ice
storage temp.   2-8°C

Description

Packaging

2 mL in glass bottle

Physical form

Solution in 0.05 M Tris buffered saline, pH 8.0, containing 0.001 mM MgCl2, 1% bovine serum albumin, and 15 mM sodium azide.

Application

Alkaline Phosphatase-Anti-Alkaline Phosphatase antibody has been used for IHC assays.

Immunohistochemistry was performed on parraffin embedded uteroplacental sections (5 um) using APAAP as the primary at a dilution of 1:100 in 0.5M TBS.

General description

Soluble complex of calf intestinal alkaline phophatase and rabbit antibodies to alkaline phosphatase.

The product may be used for immunohistology and immunocytology assays applying the Alkaline Phosphatase-Anti-Alkaline Phosphatase (APAAP) technique. This method involves the use of primary rabbit antibodies, a bridging antibody, the APAAP complex and a suitable substrate.

Price and Availability


Antibody Explorer

Explore with Confidence

Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
Protocols & Articles

Articles

How to Choose a Secondary Antibody

The following information is provided to help you decide which secondary antibody may be best for your particular application.
Keywords: Amplification, Enzyme-linked immunosorbent assay, Flow cytometry, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western blot

Secondary Antibodies, Conjugates and Kits

Secondary antibodies are polyclonal or monoclonal antibodies that bind to primary antibodies or antibody fragments, such as the Fc or Fab regions. They are typically labeled with probes that make the...
Keywords: Absorption, Adsorption, Amplification, Bacterial conjugations, Cancer, Digestions, Electrophoresis, Enzyme-linked immunosorbent assay, Flow cytometry, Gel electrophoresis, Gene expression, Hormones, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunology, Immunoprecipitation, Infrared spectroscopy, Microbiology, Microscopy, Purification, Vitamins, Western blot

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?