• USA Home
  • A9846 - 2-Aminobenzoyl-Gly(t-butyl)-Asn(methyl)2-Ala-Ser-Ser-Arg-Leu-Tyr(NO2)-Tyr-Arg trifluoroacetate salt

EMAIL THIS PAGE TO A FRIEND
A9846 Sigma

2-Aminobenzoyl-Gly(t-butyl)-Asn(methyl)2-Ala-Ser-Ser-Arg-Leu-Tyr(NO2)-Tyr-Arg trifluoroacetate salt

>94% (HPLC)

Synonym: Anthraniloyl-Gly(t-Butyl)-Asn(Methyl)2-Ala-Ser-Ser-Arg-Leu-3-nitro-Tyr-Arg

  • CAS Number 204909-37-5

  • Empirical Formula (Hill Notation) C61H97N19O18

  • Molecular Weight 1384.54

  •  MDL number MFCD03452831

Purchase

Description

Amino Acid Sequence

Abz-Gly-tBu-Asn-Me-Me-Ala-Ser-Ser-Arg-Leu-NO2-Tyr-Arg

Application

Fluorogenic substrate for the determination of human cytomegalovirus protease

Price and Availability


Detect Your Unseen Protein

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?