EMAIL THIS PAGE TO A FRIEND
A9951 Sigma

Aldehyde–Agarose

aqueous suspension

Purchase

Properties

Related Categories Activated/Functionalized Matrices, Affinity Chromatography, Molecular Biology, Protein Chromatography, Proteomics More...
form   aqueous suspension
extent of labeling   40-50 μmol per mL
matrix   cross-linked 4% beaded agarose
matrix spacer   6 atoms (when amino-ligands are coupled to the aldehyde group)
capacity   ~10 mg/g coupling capacity (BSA)
storage temp.   2-8°C

Description

Physical form

Suspension in 0.02% aqueous sodium azide

Application

Aldehyde Agarose is used in affinity chromatography. It has been used in research for the immobilization and stabilization of enzymes.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?