• USA Home
  • APREST82297 - PrEST Antigen DNASE1L2

EMAIL THIS PAGE TO A FRIEND
APREST82297 Sigma

PrEST Antigen DNASE1L2

Prestige Antigens Powered by Atlas Antibodies, buffered aqueous solution

Synonym: DNAS1L2

Purchase

Properties

Related Categories Antibodies, Prestige Antigens, Prestige Antigens C - D More...
recombinant   expressed in E. coli
assay   >80% (SDS-PAGE)
form   buffered aqueous solution
mol wt   predicted mol wt 23 kDa
purified by   immobilized metal affinity chromatography (IMAC)
concentration   ≥0.5 mg/mL
immunogen sequence   TTVGNSDCAYDRIVACGARLRRSLKPQSATVHDFQEEFGLDQTQALAISD
Ensembl | human accession no.   ENSG00000167968
UniProt accession no.   Q92874
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... DNASE1L2(1775)

Description

Application

Suitable as a blocking agent using corresponding antibodies.

General description

Recombinant protein fragment of Human DNASE1L2 with N-terminal His6ABP (ABP = Albumin Binding Protein derived from Streptococcal Protein G) fusion tag.

Linkage

Corresponding Antibody HPA044714.

Preparation Note

The protein solution should be gently mixed before use. Optimal concentrations and conditions for each application should be determined by the user.

Physical form

Solution in 1 M urea-PBS, pH 7.4

Legal Information

Prestige Antigens is a trademark of Sigma-Aldrich Co. LLC

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR 
UN 1789 8 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?