Synonym: Anti-Jumonji domain containing 3, Histone lysine demethylase, Anti-KIAA0346
| Related Categories | Alphabetical Index, Antibodies, Antibodies to Histones, Epigenetics, J, |
| species reactivity | human, mouse, rat, canine, |
| application(s) | western blot: suitable |
| clone | polyclonal |
| antibody form | IgG fraction of antiserum |
| form | lyophilized powder |
| immunogen sequence |
AGLQGVVVLEQYRTEELAQAYDAFT |
| storage temp. | −20°C |
| Gene Information | human ... JMJD3(23135) |
| NCBI accession no. |
NP_001073893 |
synthetic peptide corresponding to a region of human JMJD3 with an internal ID of P22940
Lyophilized from PBS buffer with 2% sucrose
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?