Synonym: Anti-MKI67 (FHA domain) interacting nucleolar phosphoprotein, Anti-NIFK, Anti-Nopp34
| Related Categories | Alphabetical Index, Antibodies, MF-MN, Primary Antibodies |
| species reactivity | human, mouse, pig, canine, rat, bovine |
| application(s) | western blot: suitable |
| clone | polyclonal |
| antibody form | affinity isolated antibody |
| form | lyophilized powder |
| immunogen sequence |
PSVKRYNRNRTLTQKLRMEERFKKK |
| storage temp. | −20°C |
| Gene Information | human ... MKI67IP(84365) |
| NCBI accession no. |
NP_115766 |
synthetic peptide corresponding to a region of human MKI67IP with an internal ID of S02402
Lyophilized from PBS buffer with 2% sucrose
Customers Also Viewed

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
clone PP-67, ascites fluid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?