EMAIL THIS PAGE TO A FRIEND
B0152 Sigma

(−)-Bebeerine

~95% (TLC)

Synonym: Curine

Purchase

Properties

Related Categories Biochemicals and Reagents, Luminescent Compounds/Detection, Stains and Dyes More...
InChI Key   NGZXDRGWBULKFA-VSGBNLITSA-N
assay   ~95% (TLC)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1544PSN1 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?