• USA Home
  • B0306 - Anti-Bcl-10 (AQ-18) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
B0306 Sigma

Anti-Bcl-10 (AQ-18) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Purchase

Properties

Related Categories AA - CA, Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology,
species reactivity   human, mouse, rat
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:250 using heat-retrieved, sections of rat spleen
  immunoprecipitation: 5-10 μg using lysate from 4-6x105 human THP-1/Raji cells
  microarray: suitable
  western blot: 1:2,500 using whole extract of human THP-1/Raji cells
  western blot: 1:500 using whole extract of mouse NIH3T3 fibroblasts
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt ~32 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... BCL10(8915)
mouse ... Bcl10(12042)
rat ... Bcl10(83477)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to the human Bcl-10 sequence (amino acids 216-233), with N-terminal lysine added, conjugated to KLH.

General description

Bcl-10 or CIPER is an N-terminal CARD (Caspase Recruitment Domain) containing protein mapped on to chromosome 1 at 1p22 and is localized in perinuclear region of cytoplasm and in membrane raft along with DPP4.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Application

Polyclonal anti-Bcl-10 (AQ-18) antibody produced in rabbit is suitable for immunoprecipitation at a concentration of 5-10μg by using lysate from 4-6x105 human THP-1/Raji cells and for immunohistochemistry at 1:250 working dilution using formalin-fixed, paraffin-embedded heat-retrieved, sections of rat spleen. The product is also suitable for western blot analysis at a working dilution of 1:2,500 using whole extract of human THP-1/Raji cells and at 1:500 using whole extract of mouse NIH3T3 fibroblasts. Further, anti-Bcl-10 (AQ-18) antibody has shown suitability for microarray analysis.

Biochem/physiol Actions

Bcl-10 is a cellular homolog of the equine herpesvirus-2 protein E-10 (vCLAP). It has been implicated in the regulation of apoptosis by interacting with caspase 9, enhancing procaspase 9 processing and triggering its activation when overexpressed in the cell. Deregulation of Bcl-10 expression leads to cellular oncogenesis.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  -
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?