• USA Home
  • B0430 - Ohaus® Compact portable balance
EMAIL THIS PAGE TO A FRIEND
B0430 Sigma
Ohaus® Compact portable balance
Ohaus CS 200
Purchase
Properties
Related Categories Balances and Accessories, Labware, Manufacturer Browser, Ohaus, Ohaus Compact Balances,
material   stainless steel weighing plate
feature   CE compliant
  CSA approved
mfr. no.   Ohaus CS 200
measuring range   200 g weighing capacity, precision: 0.1 g
platform diam. × diam.   145 x 133 mm × 5.75 x 5.25  in.
battery power   3 AA (alkaline batteries (included))
Description
General description
Power: 3-AA alkaline batteries (included) or AC adapter (optional); Display: LCD, Units: pounds, grams, ounces; platform: 5.25 × 4.75 in., dimensions: 7.6 × 5.3 × 1.5 in.
Features: Low battery indicator, overload and underload indicators, auto shut-off, rugged plastic construction, tactile feedback keypad.
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?