• USA Home
  • B0653 - Batteries, button cells

EMAIL THIS PAGE TO A FRIEND
B0653 Sigma

Batteries, button cells

2032, 3 V Lithium Cell

Synonym: Batteries, Eveready® button cells

Purchase

Properties

Related Categories Batteries, Eveready, Labware, Manufacturer Browser
mfr. no.   EVEREADY
battery power   3 V lithium

Description

Compatibility

Equivalent to IEC No. CR2032

Other Notes

Replacement for: 5004LC, BR2032, DL2032, ECR2032, ST-T15

Legal Information

Eveready is a registered trademark of Eveready Battery Co.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?