• USA Home
  • B0682 - Sartorius pipette tips
EMAIL THIS PAGE TO A FRIEND
B0682 Sigma
Sartorius pipette tips
volume range 0.5-300 μL, filtered tips, sterile
Purchase
Properties
material   colorless tip
sterility   sterile
feature   barrier
packaging   pack of 960
mfr. no.   Sartorius, 783207
volume range   0.5-300 μL
Description
General description
Sartorius makes pipettor tips for use with all Sartorius and most other brand pipettors providing excellent reproducibility and accuracy. All tips and trays are autoclavable. Sartorius Standard Tips of the Biohitfamily are made of premium-grade virgin polypropylene.
The tips are available in autoclavable (121°C; 20 min; 1 bar) Single Tray boxes (as certifed RNase, Dnase and endotoxin free), in space saving System and in Bulk packages. Electron beam pre-sterilized tips in trays are also available.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?