EMAIL THIS PAGE TO A FRIEND
B0754 Sigma

Boc-Pro-Pro

  • CAS Number 15401-08-8

  • Empirical Formula (Hill Notation) C15H24N2O5

  • Molecular Weight 312.36

  •  MDL number MFCD00057246

  •  PubChem Substance ID 24891533

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   XGDABHXCVGCHBB-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?