EMAIL THIS PAGE TO A FRIEND
B0759 Sigma

Bordeaux red

Synonym: Acid Red 17

  •  Colour Index Number 16180

  •  EC Number 227-489-5

Purchase

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
QJ6479500

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?