• USA Home
  • B0877 - Boron trichloride solution

EMAIL THIS PAGE TO A FRIEND
B0877 Sigma

Boron trichloride solution

10% in methanol

  • CAS Number 10294-34-5

  • Empirical Formula (Hill Notation) BCl3

  • Molecular Weight 117.17

  •  MDL number MFCD00011313

Purchase

Properties

InChI Key   FAQYAMRNWDIXMY-UHFFFAOYSA-N
concentration   10% in methanol
storage temp.   2-8°C

Description

Application

For preparing methyl esters of fatty acids and for transesterification of triglycerides.

Caution

Shelf life is extended considerably in sealed ampules.

Packaging

Sealed ampules.

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3286 8(6.1)(3) / PGII
WGK Germany 
3
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

295019 Boron trichloride, 99.9%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?