EMAIL THIS PAGE TO A FRIEND
B100 Sigma

Nucleosides

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleosides, Nucleosides, Nucleotides, Oligonucleotides More...
storage temp.   2-8°C

Description

Other Notes

4 Nucleosides, 100mg of each
A9251Adenosine
C4654 Cytidine free base, cell culture tested
G6752Guanosine
U3750Uridine

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3316 9
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?