EMAIL THIS PAGE TO A FRIEND
B1142 Sigma

Boc-D-Pro-OSu

Synonym: Boc-D-proline N-hydroxysuccinimide ester

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   DICWIJISMKZDDY-SECBINFHSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?