• USA Home
  • B1260 - N-Benzoyl-Phe-Val-Arg 4-methoxy-β-naphthylamide hydrochloride

EMAIL THIS PAGE TO A FRIEND
B1260 Sigma

N-Benzoyl-Phe-Val-Arg 4-methoxy-β-naphthylamide hydrochloride

  • CAS Number 102029-41-4

  • Empirical Formula (Hill Notation) C38H45N7O5 · HCl

  • Molecular Weight 716.27

  •  MDL number MFCD00038924

  •  PubChem Substance ID 24891574

Purchase

Description

Substrates

Substrate for thrombin.

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?