EMAIL THIS PAGE TO A FRIEND
B1278 Sigma

Benzyloxyurea

  • CAS Number 2048-50-2

  • Empirical Formula (Hill Notation) C8H10N2O2

  • Molecular Weight 166.18

  •  MDL number MFCD00047867

  •  PubChem Substance ID 24891578

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks, Ureas More...
InChI Key   IXRIVZOKBPYSPK-UHFFFAOYSA-N
assay   >99% (TLC)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?