• USA Home
  • B1301 - Bile esculin agar base

EMAIL THIS PAGE TO A FRIEND
B1301 Sigma

Bile esculin agar base

pH 6.6±0.2 (25 °C)

Purchase

Properties

pH   6.6±0.2 (25 °C)

Description

Application

Recommended for isolation and presumptive identification of Group D Streptococci.

Other Notes

Components (g/L)
Peptic digest of animal tissue 5.00
Beef extract 3.00
Oxgall 40.00
Ferric citrate 0.50
Agar 15.00

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

E8250 Esculin hydrate, ≥98%

replaced by

Product #

Description

Add to Cart

48300 Bile esculin agar, for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?