• USA Home
  • B1313 - Anti-BLVRB (122-135) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
B1313 Sigma

Anti-BLVRB (122-135) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-BVRB, Anti-FLR, Anti-Flavin reductase

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Cell Stress, Antibodies to Oxidative Stress Proteins,
species reactivity   human
application(s)   western blot: 1:500-1:2,000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt ~22 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... BLVRB(645)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 122-135 of human BLVRB

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Application

Anti-BLVRB (122-135) antibody produced in rabbit is suitable for western blotting at a working dilution of 1:500-1:2000.
Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about the Sigma Life Science and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about the Sigma Life Science and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Biochem/physiol Actions

BLVRB or Biliverdin IX β reductase (BVR-B) gene encodes a biliverdin reductase enzyme, mapped at 19q13.1-q13.2 and is widely expressed in erythrocytes and liver and to lower level in heart, lung, adrenal gland and cerebrum. It is a broad specificity bearing oxidoreductase that plays a pivotal role in heme metabolism in mammals by catalyzing the pyridine nucleotide-dependent production of bilirubin-IXβ. It is also crucial for NADPH-dependent reduction of a variety of flavins like- FAD or FMN, biliverdins, riboflavin, methemoglobin and PQQ (pyrroloquinoline quinone). Furthermore, BLVRB converts biliverdin to bilirubin, facilitates tetrapyrrole clearance in fetus. It also carries out FMN and NADPH dependent reduction of complexed Fe3+ iron to Fe2+.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?