• USA Home
  • B136 - S(−)-6-Bromo-APB hydrobromide

EMAIL THIS PAGE TO A FRIEND
B136 Sigma

S(−)-6-Bromo-APB hydrobromide

solid

  • Empirical Formula (Hill Notation) C19H20BrNO2 · HBr

  • Molecular Weight 455.18

  •  MDL number MFCD00153775

  •  PubChem Substance ID 329773180

Purchase

Properties

Related Categories Agonists, Cell Biology, Cell Signaling and Neuroscience, Dopaminergics, Neuroscience,
InChI Key   CTAUBYSSTAODOD-NTISSMGPSA-N
form   solid
color   off-white
solubility   ethanol: soluble
storage temp.   2-8°C

Description

Biochem/physiol Actions

Weak D1 dopamine receptor agonist; less potent enantiomer.

Caution

Photosensitive

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?