• USA Home
  • B1398 - Benzyl 4-O-β-D-galactopyranosyl-β-D-glucopyranoside

EMAIL THIS PAGE TO A FRIEND
B1398 Sigma

Benzyl 4-O-β-D-galactopyranosyl-β-D-glucopyranoside

Synonym: β-D-Gal-(1→4)-β-D-Glc-1→OCH2Ph, Benzyl β-lactoside

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates A-C, Disaccharide More...
InChI Key   HTGUDZYKBHFIRM-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?