• USA Home
  • B141 - (±)-4-Bromohomoibotenic acid

EMAIL THIS PAGE TO A FRIEND
B141 Sigma

(±)-4-Bromohomoibotenic acid

solid

Synonym: (±)-α-Amino-4-bromo-2,3-dihydro-3-oxo-5-isoxazolepropanoic acid

  • CAS Number 71366-32-0

  • Empirical Formula (Hill Notation) C6H7BrN2O4

  • Molecular Weight 251.03

  •  MDL number MFCD00153776

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   JRTOQOAGTSUNHA-UHFFFAOYSA-N
form   solid
color   white
solubility   H2O: moderately soluble

Description

Biochem/physiol Actions

Potent AMPA receptor agonist

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?